Merck
  • Invasive aspergillosis in patients with hematological malignancies in the Czech and Slovak republics: Fungal InfectioN Database (FIND) analysis, 2005-2009.
Invasive aspergillosis in patients with hematological malignancies in the Czech and Slovak republics: Fungal InfectioN Database (FIND) analysis, 2005-2009.
International journal of infectious diseases : IJID : official publication of the International Society for Infectious Diseases (2012-10-23)
Zdenek Racil, Barbora Weinbergerova, Iva Kocmanova, Jan Muzik, Michal Kouba, Lubos Drgona, Lucia Masarova, Tomas Guman, Elena Tothova, Kristina Forsterova, Jan Haber, Barbora Ziakova, Eva Bojtarova, Jan Vydra, Peter Mudry, Renata Foralova, Daniela Sejnova, Nada Mallatova, Vit Kandrnal, Petr Cetkovsky, Jiri Mayer
RESUMO
To evaluate risk factors, diagnostic procedures, and treatment outcomes of invasive aspergillosis (IA) in patients with hematological malignancies. A retrospective analysis of data from proven/probable IA cases that occurred from 2005 to 2009 at 10 hematology centers was performed. We identified 176 IA cases that mainly occurred in patients with acute leukemias (58.5%), mostly those on induction/re-induction treatments (39.8%). Prolonged neutropenia was the most frequent risk factor for IA (61.4%). The lungs were the most frequently affected site (93.8%) and computed tomography detected abnormalities in all episodes; however, only 53.7% of patients had findings suggestive of IA. Galactomannan (GM) detection in serum or bronchoalveolar lavage fluid (positive in 79.1% and 78.8% of episodes, respectively) played a crucial role in IA diagnosis. Neutrophil count and antifungal prophylaxis did not influence the GM positivity rate, but empirical therapy decreased this rate (in serum). Of the IA cases, 53.2% responded to initial antifungal therapy. The combination of voriconazole and echinocandin, even as initial or salvage therapy, did not perform better than voriconazole monotherapy (p=0.924 for initial therapy and p=0.205 for salvage therapy). Neutrophil recovery had a significant role in the response to initial (but not salvage) antifungal therapy. Our retrospective analysis identified key diagnostic and treatment characteristics, and this understanding could improve the management of hematological malignancy patients with IA.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
D-Galacto-D-mannan from Ceratonia siliqua, ~95% (HPLC)