Merck
Complementary and alternative medicine usage by patients of a dental school clinic.
Special care in dentistry : official publication of the American Association of Hospital Dentists, the Academy of Dentistry for the Handicapped, and the American Society for Geriatric Dentistry (2012-09-05)
Michael L Spector, Mark Fischer, Deborah V Dawson, David C Holmes, Colleen Kummet, Nicole L Nisly, Karen A K Baker
RESUMO
This pilot study investigated the prevalence and specific reasons for usage of complementary and alternative medicine (CAM) among patients of a dental school clinic. Four hundred and two patients completed a 30-page survey on CAM usage. A higher rate of CAM usage was found in this dental school clinic population than rates previously reported in a general population. More than three-quarters (76.1%) of the respondents reported using at least one CAM treatment in the past 12 months; 93.3% reported using at least one CAM treatment at some time in their lives. High rates of chiropractic use were found in this population. Tooth pain was the most frequently reported dental condition motivating CAM use. About 10% of dental school clinic patients use topical oral herbal and/or natural products to treat dental conditions, most frequently for preventive/oral health reasons or for tooth pain.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Tea tree oil, FG
Preço e disponibilidade não estão disponíveis no momento.