Merck
  • An isomorphous series of cubic, copper-based triazolyl isophthalate MOFs: linker substitution and adsorption properties.
An isomorphous series of cubic, copper-based triazolyl isophthalate MOFs: linker substitution and adsorption properties.
Inorganic chemistry (2012-07-04)
Jörg Lincke, Daniel Lässig, Merten Kobalz, Jens Bergmann, Marcel Handke, Jens Möllmer, Marcus Lange, Christian Roth, Andreas Möller, Reiner Staudt, Harald Krautscheid
RESUMO
An isomorphous series of 10 microporous copper-based metal-organic frameworks (MOFs) with the general formulas (∞)(3)[{Cu(3)(μ(3)-OH)(X)}(4){Cu(2)(H(2)O)(2)}(3)(H-R-trz-ia)(12)] (R = H, CH(3), Ph; X(2-) = SO(4)(2-), SeO(4)(2-), 2 NO(3)(2-) (1-8)) and (∞)(3)[{Cu(3)(μ(3)-OH)(X)}(8){Cu(2)(H(2)O)(2)}(6)(H-3py-trz-ia)(24)Cu(6)]X(3) (R = 3py; X(2-) = SO(4)(2-), SeO(4)(2-) (9, 10)) is presented together with the closely related compounds (∞)(3)[Cu(6)(μ(4)-O)(μ(3)-OH)(2)(H-Metrz-ia)(4)][Cu(H(2)O)(6)](NO(3))(2)·10H(2)O (11) and (∞)(3)[Cu(2)(H-3py-trz-ia)(2)(H(2)O)(3)] (12(Cu)), which are obtained under similar reaction conditions. The porosity of the series of cubic MOFs with twf-d topology reaches up to 66%. While the diameters of the spherical pores remain unaffected, adsorption measurements show that the pore volume can be fine-tuned by the substituents of the triazolyl isophthalate ligand and choice of the respective copper salt, that is, copper sulfate, selenate, or nitrate.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Isophthalic acid, 99%