- Enantioselective Michael addition of 3-aryl-substituted oxindoles to methyl vinyl ketone catalyzed by a binaphthyl-modified bifunctional organocatalyst.
Enantioselective Michael addition of 3-aryl-substituted oxindoles to methyl vinyl ketone catalyzed by a binaphthyl-modified bifunctional organocatalyst.
Molecules (Basel, Switzerland) (2012-06-20)
Hyun Joo Lee, Saet Byeol Woo, Dae Young Kim
PMID22710826
RESUMO
The enantioselective conjugate addition reaction of 3-aryl-substituted oxindoles with methyl vinyl ketone promoted by binaphthyl-modified bifunctional organocatalysts was investigated. The corresponding Michael adducts, containing a quaternary center at the C3-position of the oxindoles, were generally obtained in high yields with excellent enantioselectivities (up to 91% ee).