Merck
  • Rapid repeat pregnancy in adolescents: do immediate postpartum contraceptive implants make a difference?
Rapid repeat pregnancy in adolescents: do immediate postpartum contraceptive implants make a difference?
American journal of obstetrics and gynecology (2012-05-29)
Kristina M Tocce, Jeanelle L Sheeder, Stephanie B Teal
RESUMO
The purpose of this study was to determine contraceptive continuation and repeat pregnancy rates in adolescents who are offered immediate postpartum etonogestrel implant (IPI) insertion. Participants in an adolescent prenatal-postnatal program were enrolled in a prospective observational study of IPI insertion (IPI group, 171) vs other methods (control group, 225). Contraceptive continuation and repeat pregnancies were determined. Implant continuation at 6 months was 96.9% (156/161 participants); at 12 months, the continuation rate was 86.3% (132/153 participants). At 6 months, 9.9% of the control participants were pregnant (21/213); there were no IPI pregnancies. By 12 months, 18.6% of control participants (38/204) experienced pregnancy vs 2.6% of IPI recipients (4/153; relative risk, 5.0; 95% confidence interval [CI], 1.9-12.7). Repeat pregnancy at 12 months was predicted by not receiving IPI insertion (odds ratio, 8.0; 95% CI, 2.8-23.0) and having >1 child (odds ratio, 2.1; 95% CI, 1.1-4.3; P = .03). IPI placement in adolescents has excellent continuation 1 year after delivery; rapid repeat pregnancy is significantly decreased compared with control participants.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Etonogestrel, ≥98% (HPLC)