Merck
  • Hydrogen evolution across nano-Schottky junctions at carbon supported MoS2 catalysts in biphasic liquid systems.
Hydrogen evolution across nano-Schottky junctions at carbon supported MoS2 catalysts in biphasic liquid systems.
Chemical communications (Cambridge, England) (2012-05-26)
Peiyu Ge, Micheál D Scanlon, Pekka Peljo, Xiaojun Bian, Heron Vubrel, Arlene O'Neill, Jonathan N Coleman, Marco Cantoni, Xile Hu, Kyösti Kontturi, Baohong Liu, Hubert H Girault
RESUMO
The activities of a series of MoS(2)-based hydrogen evolution catalysts were studied by biphasic reactions monitored by UV/Vis spectroscopy. Carbon supported MoS(2) catalysts performed best due to an abundance of catalytic edge sites and strong electronic coupling of catalyst to support.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Molybdenum(IV) sulfide, powder
Sigma-Aldrich
Molybdenum(IV) sulfide, powder, <2 μm, 98%