Merck
  • Nitric oxide and guanylate cyclase signalling are differentially involved in gonadotrophin (LH) release responses to two endogenous GnRHs from goldfish pituitary cells.
Nitric oxide and guanylate cyclase signalling are differentially involved in gonadotrophin (LH) release responses to two endogenous GnRHs from goldfish pituitary cells.
Journal of neuroendocrinology (2012-04-11)
A N Meints, J G Pemberton, J P Chang
RESUMO
Nitric oxide synthase (NOS) immunoreactivity is present in goldfish gonadotrophs. The present study investigated whether two native goldfish gonadotrophin-releasing hormones (GnRHs), salmon (s)GnRH and chicken (c)GnRH-II, use NOS/nitric oxide (NO) and soluble guanylate cyclase (sGC)/cyclic (c)GMP/protein kinase G (PKG) signalling to stimulate maturational gonadotrophin [teleost gonadotrophin-II, luteinising hormone (LH)] release. In cell column perifusion experiments with dispersed goldfish pituitary cells, the application of three NOS inhibitors (aminoguanidine hemisulphate, 1400W and 7-nitroindazole) and two NO scavengers [2-phenyl-4,4,5,5-tetramethylimidazoline-1-oxyl-3-oxide (PTIO) and rutin hydrate] reduced sGnRH-elicited, but not cGnRH-II-induced, LH increases. The NO donor sodium nitroprusside (SNP) increased NO production in goldfish pituitary cells in static incubation. SNP-stimulated LH release in column perifusion was attenuated by PTIO and the sGC inhibitor 1H-(1,2,4)oxadiazolo[4,3-a]quinoxalin-1-oneon (ODQ), and additive to responses elicited by cGnRH-II, but not sGnRH. ODQ and the PKG inhibitor KT5823 decreased sGnRH- and cGnRH-II-stimulated LH release. Similarly, the LH response to dibutyryl cGMP was reduced by KT5823. These results indicate that, although only sGnRH uses the NOS/NO pathway to stimulate LH release, both GnRHs utilise sGC/PKG to increase LH secretion.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
KT 5823, ≥85% (HPLC), lyophilized powder