Merck
Legionella pneumophila lung abscess associated with immune suppression.
Internal medicine journal (2012-03-23)
S D Guy, L J Worth, K A Thursky, P A Francis, M A Slavin
RESUMO
Legionella species are a common cause of community-acquired pneumonia, infrequently complicated by cavitary disease. We describe Legionella pneumophila pneumonia and abscess formation in an immunosuppressed patient receiving corticosteroid therapy for metastatic breast carcinoma. The predisposing role of corticosteroids is discussed and the management of this complication is reviewed.
MATERIAIS
Número do produto
Marca
Descrição do produto
Roxithromycin, European Pharmacopoeia (EP) Reference Standard