Merck
The colorimetric detection and quantitation of total protein.
Current protocols in cell biology (2011-09-08)
Randall I Krohn
RESUMO
Protein quantification is an important step for handling protein samples for isolation and characterization; it is a prerequisite step before submitting proteins for chromatographic, electrophoretic, or immunochemical analysis and separation. Colorimetric methods are fast, simple, and not laborious. This unit describes a number of assays able to detect protein concentrations in the low microgram to milligram per milliliter ranges in a variety of formats.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
2,2′-Biquinoline-4,4′-dicarboxylic acid, ≥90% (TLC)