Merck
  • Photodynamic treatment of Chaoborus crystallinus larvae with chlorophyllin induces necrosis and apoptosis.
Photodynamic treatment of Chaoborus crystallinus larvae with chlorophyllin induces necrosis and apoptosis.
Photochemistry and photobiology (2011-06-28)
Stephanie Wohllebe, Claudia Ulbrich, Daniela Grimm, Jessica Pietsch, Gilmar Erzinger, Roland Richter, Michael Lebert, Peter Rolf Richter, Donat-Peter Häder
RESUMO
Chlorophyllin kills mosquito larvae (Culex, Aedes) in the aquatic habitat at low concentrations via photodynamic reactions under irradiation. The effects of chlorophyllin were investigated at the cellular level using the transparent larvae of Chaoborus crystallinus as a model system. Their transparency enabled in situ fluorescence investigation, showing that chlorophyllin accumulates in the intestine of the larvae. Uptake of chlorophyllin at room temperature took about 2 h. The fluorescence signal peaked after 5 h of incubation. Chlorophyllin accumulates up to about 15 ng per larvae. The intestine of treated larvae was dissected and stained with several dyes (acridine orange, Hoechst 33342 and propidium iodide). Apoptosis and necrosis increased with higher concentrations of chlorophyllin (to a smaller extent in dark controls) and were elevated in irradiated samples. Single cells from treated larvae were isolated and subjected to Annexin V flow cytometry. The fraction of apoptotic and necrotic cells increased significantly at a high chlorophyllin concentration (21.4 mg L(-1)) and under intensive irradiation. The activity of caspases-3, -8 and -9 as well as Bcl-2 and cytochrome c was investigated by means of western blot analysis. The data suggest a possible chlorophyllin concentration-dependent shift of the apoptotic pathway.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Chlorophyllin sodium copper salt, commercial grade