Merck
  • The dithiothreitol-stimulated dissociation of the chloroplast coupling factor 1 epsilon-subunit is reversible.
The dithiothreitol-stimulated dissociation of the chloroplast coupling factor 1 epsilon-subunit is reversible.
Biochimica et biophysica acta (1990-05-15)
R J Duhe, B R Selman
RESUMO
The chloroplast coupling factor 1 complex (CF1) contains an epsilon-subunit which inhibits the CF1 ATPase activity. Chloroform treatment of Chlamydomonas reinhardtii thylakoid membranes solubilizes only forms of the enzyme which apparently lack the delta-subunit. Four interrelated observations are described in this paper. (1) The dithiothreitol- (DTT) induced ATPase activation of CF1(-delta) and the DTT-induced formation of a physically resolvable CF1(-delta,epsilon) from the CF1(-delta) precursor are compared. The similar time-courses of these two phenomena suggest that the dissociation of the epsilon-subunit is an obligatory process in the DTT-induced ATPase activation of soluble CF1. (2) The reversible dissociation of the epsilon-subunit of the CF1 is demonstrated by the exchange of subunits between distinguishable oligomers. 35S-labelled chloroplast coupling factor 1 lacking the delta and epsilon subunits [CF1(-delta,epsilon)] was added to a solution of non-radioactive coupling factor 1 lacking only the delta subunit [CF1(-delta)]. After separation of the two enzyme forms, via high resolution anion-exchange chromatography, radioactivity was detected in the chromatographic fractions containing CF1(-delta). (3) epsilon-deficient CF1 can be resolved from DTT pretreated epsilon-containing CF1 for several days after the removal of DTT. On the other hand, brief incubation of the DTT pretreated epsilon-containing CF1 with low concentrations of o-iodosobenzoate results in chromatographs containing only the peak of epsilon-containing CF1. A simple explanation for this phenomenon is that reduction of CF1 with DTT increases the apparent dissociation constant for the epsilon-subunit to an estimated 3.5 x 10(-8) M (+/- 1.0 x 10(-8) M) from a value of less than or equal to 5 x 10(-11) M for the oxidized enzyme. (4) ATPase activity data show that oxidation of the epsilon-deficient enzyme does not completely inhibit its manifest activity, but oxidation of DTT pre-treated CF1 which contains the epsilon-subunit completely inhibits manifest activity. A simple model is proposed for the influence of the oxidation state of the soluble enzyme on the distribution of ATPase-inactive and ATPase-active subunit configurations.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
2-Iodobenzoic acid, 98%