- In vitro evaluation of 5-arylidene-2-thioxo-4-thiazolidinones active as aldose reductase inhibitors.
In vitro evaluation of 5-arylidene-2-thioxo-4-thiazolidinones active as aldose reductase inhibitors.
Bioorganic & medicinal chemistry letters (2010-12-07)
Rosanna Maccari, Antonella Del Corso, Marco Giglio, Roberta Moschini, Umberto Mura, Rosaria Ottanà
PMID21129963
RESUMO
2-Thioxo-4-thiazolidinone derivatives were evaluated as aldose reductase inhibitors (ARIs) and most of them exhibited good or excellent in vitro efficacy. Out of the tested compounds, most N-unsubstituted analogues were found to possess inhibitory effects at low micromolar doses and two of them exhibited higher potency than sorbinil, used as a reference drug. The insertion of an acetic chain on N-3 of the thiazolidinone scaffold led to analogues with submicromolar affinity for ALR2 and IC(50) values very similar to that of epalrestat, the only ARI currently used in therapy.
MATERIAIS