- Palladium(II) complexes with R(2)edda derived ligands. Part IV. O,O'-dialkyl esters of (S,S)-ethylenediamine-N,N'-di-2-(4-methyl)-pentanoic acid dihydrochloride and their palladium(II) complexes: synthesis, characterization and in vitro antitumoral activity against chronic lymphocytic leukemia (CLL) cells.
Palladium(II) complexes with R(2)edda derived ligands. Part IV. O,O'-dialkyl esters of (S,S)-ethylenediamine-N,N'-di-2-(4-methyl)-pentanoic acid dihydrochloride and their palladium(II) complexes: synthesis, characterization and in vitro antitumoral activity against chronic lymphocytic leukemia (CLL) cells.
European journal of medicinal chemistry (2010-06-24)
Jelena M Vujić, Milica Cvijović, Goran N Kaluderović, Marija Milovanović, Bojana B Zmejkovski, Vladislav Volarević, Nebojsa Arsenijević, Tibor J Sabo, Srećko R Trifunović
PMID20570025
RESUMO
Four novel bidentate N,N'-ligand precursors, including O,O'-dialkyl esters (alkyl = ethyl, n-propyl, n-butyl and n-pentyl), L1 x 2 HCl-L4 x 2 HCl, of (S,S)-ethylenediamine-N,N'-di-2-(4-methyl)-pentanoic acid dihydrochloride [(S,S)-H(4)eddl]Cl(2) and the corresponding palladium(II) complexes 1-4, were prepared and characterized by IR, (1)H NMR and (13)C NMR spectroscopy and elemental analysis. In vitro cytotoxicity of all compounds was determined against chronic lymphocytic leukemia cells (CLL). The compounds were found to exhibit higher antitumoral activity than cisplatin. The most active compound 2, [PdCl(2){(S,S)-nPr(2)eddl}], was found to be 13.6 times more active than cisplatin on CLL cells.
MATERIAIS
Número do produto
Marca
Descrição do produto