Merck
  • Electrical and optical properties of copper and nickel molecular materials with tetrabenzo [b,f,j,n] [1,5,9,13] tetraazacyclohexadecine thin films grown by the vacuum thermal evaporation technique.
Electrical and optical properties of copper and nickel molecular materials with tetrabenzo [b,f,j,n] [1,5,9,13] tetraazacyclohexadecine thin films grown by the vacuum thermal evaporation technique.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2009-12-08)
A Rodriguez, M E Sánchez-Vergara, V García-Montalvo, A Ortiz-Rebollo, J R Alvarez-Bada, C Alvarez-Toledano
RESUMO
Semiconducting molecular-material thin-films of tetrabenzo (b,f,j,n) [1,5,9,13] tetraazacyclohexadecine copper(II) and nickel(II) bisanthraflavates have been prepared by using vacuum thermal evaporation on Corning glass substrates and crystalline silicon wafers. The films thus obtained were characterized by infrared spectroscopy (FTIR), atomic force microscopy (AFM), ultraviolet-visible (UV-vis) spectroscopy and ellipsometry. IR spectroscopy showed that the molecular-material thin-films exhibit the same intra-molecular bonds as the original compounds, which suggests that the thermal evaporation process does not significantly alter their bonds. The optical band-gap values calculated from the absorption coefficient may be related to non-direct electronic interband transitions. The effect of temperature on conductivity was also measured in these samples. It was found that the temperature-dependent electric current is always higher for the nickel-based material and suggests a semiconductor-like behavior with conductivities in the order of 10(-8)Omega(-1)cm(-1).
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Anthraflavic acid, technical grade, 90%