Merck
  • Isolation of dieldrin- and endrin-degrading bacteria using 1,2-epoxycyclohexane as a structural analog of both compounds.
Isolation of dieldrin- and endrin-degrading bacteria using 1,2-epoxycyclohexane as a structural analog of both compounds.
Applied microbiology and biotechnology (2008-09-05)
Emiko Matsumoto, Youhei Kawanaka, Sun-Ja Yun, Hiroshi Oyaizu
RESUMO
This report describes the selective isolation of dieldrin- and endrin-degrading bacteria from soil with high degradation activity toward dieldrin and endrin. Several enrichment cultures from the soil were arranged with several structural analogs of dieldrin and endrin as a growth substrate and examined for their degradation activities toward dieldrin and endrin. An enrichment culture with 1,2-epoxycyclohexane (ECH) was found to aerobically degrade dieldrin and endrin. Denaturing gradient gel electrophoresis (DGGE) indicated that three types of bacteria were predominant in the ECH enrichment culture. Of the three major bacteria, two isolates, Burkholderia sp. strain MED-7 and Cupriavidus sp. strain MED-5, showed high degradation activity toward dieldrin and endrin. The degradation efficiencies of strain MED-7 and MED-5 were 49% and 38% toward dieldrin, respectively, and 51% and 40% toward endrin, respectively, in the presence of ECH for 14 days. These results indicated that ECH was a useful substrate for selective and efficient isolation of dieldrin- and endrin-degrading bacteria from soil containing numerous bacteria. Interestingly, the two isolates could also degrade dieldrin and endrin even in the absence of ECH. These are the first microorganisms demonstrated to grow on dieldrin and endrin as the sole carbon and energy source under aerobic conditions.