Merck
  • Self-assembly of perfunctionalized beta-cyclodextrins on a quartz crystal microbalance for real-time chiral recognition.
Self-assembly of perfunctionalized beta-cyclodextrins on a quartz crystal microbalance for real-time chiral recognition.
Langmuir : the ACS journal of surfaces and colloids (2008-07-12)
Changhua Xu, Siu Choon Ng, Hardy Sze On Chan
RESUMO
This study reports the development of novel chiral sensors based on the self-assembly of perfunctionalized beta-cyclodextrins (beta-CD) on a quartz crystal microbalance transducer for real time chiral recognition. Ten chiral sensors immobilized with mercaptyl perfunctionalized beta-cyclodextrins were found to exhibit promising enantioselectivity in the gas phase. Well-defined sizes of molecular cavities of the modified beta-CDs associated with cooperative weak interactions with the host molecules afforded enhanced chiral discrimination. This study contributes to the realization of novel chiral sensors applicable for real-time recognition and analysis of enantiomeric alcohols and lactates.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
(−)-Methyl L-lactate, 98%, optical purity ee: 97% (GLC)
Preço e disponibilidade não estão disponíveis no momento.
Sigma-Aldrich
(+)-Methyl D-lactate, 98%, optical purity ee: 96% (GLC)
Preço e disponibilidade não estão disponíveis no momento.
Sigma-Aldrich
Methyl DL-lactate, ≥97.0% (GC)
Preço e disponibilidade não estão disponíveis no momento.