Merck
  • Quantitative investigation of the role of breast cancer resistance protein (Bcrp/Abcg2) in limiting brain and testis penetration of xenobiotic compounds.
Quantitative investigation of the role of breast cancer resistance protein (Bcrp/Abcg2) in limiting brain and testis penetration of xenobiotic compounds.
Drug metabolism and disposition: the biological fate of chemicals (2008-03-07)
Junichi Enokizono, Hiroyuki Kusuhara, Atsushi Ose, Alfred H Schinkel, Yuichi Sugiyama
RESUMO
The role of breast cancer resistance protein (BCRP/ABCG2) in limiting the brain and testis penetration of xenobiotic compounds in the blood-brain and -testis barriers was investigated using Bcrp(-/-) mice. Tissue/plasma concentration ratios in the brain (K(p,brain)) and testis (K(p,testis)) obtained under steady-state conditions were significantly increased in Bcrp(-/-) mice for PhIP (2-amino-1-methyl-6-phenylimidazo[4,5-b]pyridine), N-hydroxyl PhIP, MeIQx (2-amino-3,8-dimethylimidazo[4,5-f]quinoxaline), dantrolene, and prazosin. In addition, the K(p,brain) of triamterene and the K(p,testis) of 4'-hydroxyl PhIP were also significantly increased in Bcrp(-/-) mice. The effect of functional impairment of Bcrp on the brain uptake of PhIP, dantrolene, and daidzein in Bcrp(-/-) mice determined using in situ brain perfusion was weaker than that observed on the K(p) values. In vitro transcellular transport experiments using cell lines expressing mouse Bcrp or P-glycoprotein (Mdr1a/Abcb1a) showed that, among the tested Bcrp substrates, PhIP, MeIQx, prazosin, and triamterene are common substrates of Bcrp and P-glycoprotein. The K(p) values of common substrates exhibited a smaller increase both in the brain and testis of Bcrp(-/-) mice than expected from the in vitro Bcrp activities. The Bcrp-specific substrates were weak acids, whereas basic or neutral BCRP substrates were also P-glycoprotein substrates. These results suggest that BCRP limits the tissue penetration of xenobiotic compounds in the blood-brain and -testis barriers, but its in vivo importance is also modulated by P-glycoprotein activity.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Triamterene, ≥99%