Merck
[Synthesis and molecule recognition capability of corilagin-molecularly imprinted polymer].
Yao xue xue bao = Acta pharmaceutica Sinica (2008-02-28)
Xiao-hong Yuan, Xiao-jie Xu, Xiao-hui Qiu
RESUMO
To study the molecule recognition capability of corilagin-molecularly imprinted polymer (MIP) by the high performance liquid chromatography (HPLC), the molecularly imprinted polymer was synthesized by using corilagin as the template. Chromatographic performance of corilagin was investigated in different mobile phases. The MIP was investigated for the recognition of corilagin and its derivatives and other compounds in the same mobile phase. The MIP exhibited very high affinity for corilagin in the mobile phase of acetonitrile. The K' value will be reduced when the content of polar solvent increased in the mobile phase. The MIP has good selectivity in the mobile phase of acetonitrile-methanol (95:5), but it has no affinity for corilagin's derivatives. The corilagin-MIP has good selectivity for corilagin and it can be used in extracting corilagin and its analogs from herbs.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Corilagin
Supelco
Corilagin, analytical standard