Merck
  • DDT- and DDE-induced disruption of ovarian steroidogenesis in prepubertal porcine ovarian follicles: a possible interaction with the main steroidogenic enzymes and estrogen receptor beta.
DDT- and DDE-induced disruption of ovarian steroidogenesis in prepubertal porcine ovarian follicles: a possible interaction with the main steroidogenic enzymes and estrogen receptor beta.
Journal of physiology and pharmacology : an official journal of the Polish Physiological Society (2008-01-16)
A K Wójtowicz, M Kajta, E Ł Gregoraszczuk
RESUMO
We evaluated impact of DDT isomers, o, p'- DDT [1, 1-dichloro-2, 2-bis (p, p'-chlorophenyl) ethylene] and p, p'-DDT [1, 1, 1-trichloro-2, 2-bis (p-chlorophenyl) ethane], and their metabolites, o, p'-DDE and p, p'-DDE, on ovarian steroidogenesis. All these compounds, except for p, p'-DDT, demonstrated estrogenic effects on steroid secretion in co-cultures of porcine prepubertal granulosa and theca cells. p,p'-DDT decreased progesterone and estradiol release, which was reversed by the addition of testosterone. In contrast, o, p'-DDT inhibited progesterone secretion with parallel stimulation of basal and testosterone-stimulated estradiol release. DDEs stimulated progesterone and estradiol secretion. The fluorometric assay confirmed that p,p'-DDE, o,p'-DDT, and o,p'-DDE stimulated aromatase activity. Western blots indicated that o,p-DDT and o,p'-DDE diminished the expression of estrogen receptor beta (ERbeta). This study demonstrated the isomer-dependent action of DDT in pig ovarian cells. We propose that DDT could disrupt ovarian steroidogenesis either by interfering with main steroidogenic enzymes or affecting ERbeta.
MATERIAIS
Número do produto
Marca
Descrição do produto
Supelco
2,4′-DDE solution, 100 μg/mL in methanol, PESTANAL®, analytical standard