Merck
  • Anticlastogenic and antigenotoxic effects of selenomethionine on doxorubicin-induced damage in vitro in human lymphocytes.
Anticlastogenic and antigenotoxic effects of selenomethionine on doxorubicin-induced damage in vitro in human lymphocytes.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2007-10-27)
Raquel Alves Dos Santos, Catarina Satie Takahashi
RESUMO
The use of antioxidants during chemotherapy has been shown to reduce or prevent the undesirable effects experienced by healthy cells. Micronutrient selenium is well known for its antioxidant properties; however, selenium exhibits a bimodal nature in that both its beneficial and toxic properties lie within a limited and narrow dose range. The present study investigated the possible protective effects of selenomethionine (SM) on the cytotoxicity, genotoxicity and clastogenicity of the chemotherapic doxorubicin (DXR), a key chemotherapic used in cancer treatment. Human peripheral lymphocytes were treated in vitro with varying concentrations of SM (0.25 microM, 0.5 microM, 1.0 microM and 2.0 microM), tested in combination with DXR (0.15 microg/mL). SM alone was not cytotoxic and when combined with DXR treatment, reduced the DNA damage index significantly, the frequency of chromosomal aberrations, the number of aberrant metaphases and the frequency of apoptotic cells. The mechanism of chemoprotection of SM may be related to its antioxidant properties as well as its ability to interfere with DNA repair pathways. Therefore this study showed that SM is effective in reducing the genetic damage induced by the antitumoral agent DXR.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Antipain, >50000 U/mg