Merck
Synthesis of santiagonamine.
Organic letters (2007-07-31)
Michael D Markey, Ying Fu, T Ross Kelly
RESUMO
The first total synthesis of santiagonamine (1) is achieved in 12 steps from isovanillin. A palladium-catalyzed Ullmann cross-coupling reaction and a photocyclization are the key steps in the synthesis.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
3-Hydroxy-4-methoxybenzaldehyde, 99%
Sigma-Aldrich
Isovanillin, ≥95.0%