Merck
GnRH analogs: options for endometriosis-associated pain treatment.
Journal of minimally invasive gynecology (2006-11-14)
Frances R Batzer
RESUMO
While none of the currently available treatment options for endometriosis pain resolved the underlying disease process, there are growing numbers of medical alternatives available. Medical options include the GnRH agonists and antagonists. Review of these treatments in the management of endometriosis pain and the insight often to the etiology of endometriosis are presented for discussion.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Nafarelin acetate salt hydrate, ≥95% (HPLC), solid
Preço e disponibilidade não estão disponíveis no momento.