Merck
Asymmetric [2 + 2] cycloaddition: total synthesis of (-)-swainsonine and (+)-6-epicastanospermine.
Organic letters (2006-10-06)
Julien Ceccon, Andrew E Greene, Jean-François Poisson
RESUMO
[reaction: see text] An asymmetric total synthesis of (-)-swainsonine and (+)-6-epicastanospermine is described from a common intermediate, which is obtained through diastereoselective [2 + 2] cycloaddition of dichloroketene to a chiral enol ether.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
(1S,6S,7R,8R,8aR)-1,6,7,8-Tetrahydroxyindolizidine, 98%