Merck
Biological markers of exposure to benzene: S-phenylcysteine in albumin.
Carcinogenesis (1992-07-01)
W E Bechtold, J K Willis, J D Sun, W C Griffith, T V Reddy
RESUMO
Results of experiments in our laboratory have shown that benzene is metabolized by animals in part to an intermediate that binds to cysteine groups in hemoglobin to form the adduct S-phenylcysteine (SPC). These results suggested that SPC in hemoglobin may be an effective biological marker for exposure to benzene. However, we could not detect SPC in the globin of humans occupationally exposed to benzene concentrations as high as 28 p.p.m. for 8 h/day, 5 days/week. As another approach, we examined the binding of benzene to cysteine groups of a different blood protein, albumin. To facilitate the process, a new method for the precipitative isolation of albumin from plasma was also developed. The isolated albumin was analyzed for SPC by isotope dilution GC-MS. We used this approach to measure SPC in the albumin of F344/N rats exposed by gavage to 0-10,000 mumol/kg benzene. Amounts of albumin-associated SPC increased as a function of dose, followed by a leveling off in the amount of SPC seen at doses greater than 1000 mumol/kg. Levels of SPC were measured in humans occupationally exposed to average concentrations of 0, 4.4, 8.4 and 23 p.p.m. benzene 8 h/day, 5 days/week. Of nine controls, seven had levels of SPC below the limit of detection (0.1 pmol SPC/mg albumin). SPC increased in the exposed groups linearly, giving a statistically significant slope (P less than 0.001) of 0.044 +/- 0.008 pmol/mg albumin/p.p.m. with an intercept of 0.135 +/- 0.095 pmol/mg albumin. From this study, we conclude that SPC in albumin may prove useful as a biomarker for benzene exposure.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
S-Phenyl-L-cysteine, 97%