Merck
  • Stereochemistry of reactions of the inhibitor/substrates L- and D-beta-chloroalanine with beta-mercaptoethanol catalysed by L-aspartate aminotransferase and D-amino acid aminotransferase respectively.
Stereochemistry of reactions of the inhibitor/substrates L- and D-beta-chloroalanine with beta-mercaptoethanol catalysed by L-aspartate aminotransferase and D-amino acid aminotransferase respectively.
Organic & biomolecular chemistry (2005-09-01)
Benjamin Adams, Kreingkrai Lowpetch, Faye Thorndycroft, Sheena M Whyte, Douglas W Young
RESUMO
Two members of the alpha-family of PLP-dependent enzymes, L-aspartate aminotransferase and D-amino acid aminotransferase, have been shown to catalyse beta-substitution of L- and D-beta-chloroalanine respectively with beta-mercaptoethanol, reactions typical of the beta-family of PLP-dependent enzymes. The reaction catalysed by L-aspartate aminotransferase has been shown to occur with retention of stereochemistry, a typical outcome for reactions catalysed by beta-family enzymes. There are also indications that the reaction catalysed by D-amino acid aminotransferase may involve retention of stereochemistry. Both enzymes have been shown to catalyse exchange at C-3 when the appropriate enantiomer of beta-chloroalanine is the substrate.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
β-Chloro-L-alanine hydrochloride, Alanine aminotransferase inhibitor
Sigma-Aldrich
β-Chloro-D-alanine hydrochloride