Merck
  • In vitro activity of ramoplanin against Clostridium difficile, including strains with reduced susceptibility to vancomycin or with resistance to metronidazole.
In vitro activity of ramoplanin against Clostridium difficile, including strains with reduced susceptibility to vancomycin or with resistance to metronidazole.
Antimicrobial agents and chemotherapy (2005-02-25)
T Peláez, L Alcalá, R Alonso, A Martín-López, V García-Arias, M Marín, E Bouza
RESUMO
We evaluated the in vitro activity of ramoplanin, an antimicrobial compound that inhibits cell wall synthesis by acting at the level of lipid intermediate formation, against Clostridium difficile. We included strains with reduced susceptibilities to vancomycin (vancomycin-intermediate [Van(i)] strains) or with resistance to metronidazole (Mtz(r)), in order to assess the potential utility of ramoplanin for the treatment of C. difficile-associated diarrhea. We tested the activity of ramoplanin against a total of 105 nonduplicate clinical isolates of toxigenic C. difficile, including 8 Van(i) isolates and 6 Mtz(r) isolates, obtained from our laboratory. Ramoplanin was active against all strains tested at concentrations ranging from 0.03 to 0.5 microg/ml (MICs at which 50 and 90% of isolates were inhibited, 0.25 microg/ml; geometric mean MIC, 0.22 microg/ml). All isolates, independently of their levels of susceptibility to vancomycin or metronidazole, were considered susceptible to ramoplanin (MICs, < or =0.5 microg/ml).
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Ramoplanin