Merck
  • Alteration of the pharmacokinetics of theophylline by rutaecarpine, an alkaloid of the medicinal herb Evodia rutaecarpa, in rats.
Alteration of the pharmacokinetics of theophylline by rutaecarpine, an alkaloid of the medicinal herb Evodia rutaecarpa, in rats.
The Journal of pharmacy and pharmacology (2005-02-22)
Yune-Fang Ueng, Tung-Hu Tsai, Ming-Jaw Don, Ruei-Ming Chen, Ta-Liang Chen
RESUMO
Rutaecarpine is a main active alkaloid present in the medicinal herb, Evodia rutaecarpa. The cytochrome P450 (CYP) 1A2 substrate, theophylline, is an important therapeutic agent for the treatment of asthma, but has a narrow therapeutic index. To evaluate the pharmacokinetic interaction of theophylline with rutaecarpine, the effects of rutaecarpine on CYP1A2 activity and theophylline pharmacokinetics were investigated. Oral treatment of Sprague-Dawley rats with 50 mg kg(-1) rutaecarpine for three days through a gastrogavage caused a 4- and 3-fold increase in liver microsomal 7-ethoxyresorufin O-deethylation (EROD) and 7-methoxyresorufin O-demethylation activity, respectively. In the kidney, rutaecarpine treatment caused a 3-fold increase in EROD activity. In the lungs, EROD activity was elevated from an undetectable to a detectable level by rutaecarpine. Pharmacokinetic parameters of theophylline were determined using a microdialysis sampling method. Rutaecarpine pre-treatment increased the clearance of theophylline in a dose-dependent manner. Pre-treatment of rats with 50 mg kg(-1) rutaecarpine caused a 3-fold increase in theophylline clearance and a 70%, 68% and 68% decrease in the area under the concentration-time curve (AUC), mean residence time (MRT) and half-life, respectively. These results demonstrated that rutaecarpine treatment elevated CYP1A2 catalytic activity and theophylline excretion in rats. In patients taking theophylline, adverse effects might be noticed when a rutaecarpine-containing herbal preparation is used concomitantly.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Resorufin methyl ether