Merck
[The comparison of luminescence properties for Ce3+, Tb3+ ions in the fluorides].
Guang pu xue yu guang pu fen xi = Guang pu (2003-08-30)
G Wu, R Chen, Q Zhang
RESUMO
The matrix K3AlF6 was prepared with a reaction of mixture of solution K2CO3 upon certain solution of Al(OH)3 plus HF (40%), dehydrating at 100 degrees C and annealing for 48 h at 600 degrees C. A series of phosphors for Ce3+, Tb3+ ions in complex fluorides KBF4 and K3AlF6 were synthesized by a solid reaction method. Their excitation and emission spectra were determined, compared with phosphors KAlF4: Ce, Tb, CaF2: Ce, Tb and AlF3: Ce, Tb. The results indicated that Ce3+ could only effectively sensitize Tb3+ in KAlF4 and enhance the emission of Tb3+ significantly. Ce3+ can not effectvely transfer energy to Tb3+ in matrixes CaF2, AlF3, KBF4 and K3AlF6.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Cerium(III) fluoride, anhydrous, powder, 99.99% trace metals basis