Merck
Extracellular purines from cells of seminiferous tubules.
Molecular and cellular biochemistry (2003-04-24)
Daniel Pens Gelain, Luiz Fernando de Souza, Elena Aida Bernard
RESUMO
It has been long postulated that extracellular purines can modulate the function of the male reproductive system by interacting with different purinergic receptors of Sertoli and germinative cells. Many authors have described the biological changes induced by extracellular ATP and/or adenosine in these cells, and some hypothetical models for paracrine communication mediated by purines were proposed; however, the cellular source(s) of these molecules in seminiferous tubules remains unknown. In this study, we demonstrated for the first time that Sertoli cells are able to release ATP (0.3 nmol/mg protein) and adenosine (0.1 nmol/mg protein) in the extracellular medium, while germinative and myoid peritubular cells are able to secrete adenosine (0.02 and 0.37 nmol/mg protein, respectively). Indeed, all the three types of cells were able to release inosine at significant concentrations (about 0.4 nmol/mg protein). This differential secretion depending on the cellular type suggests that these molecules may be involved in the paracrine regulation and/or control of the maturation processes of these cells.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Adenosine 5′-(α,β-methylene)diphosphate, ADP analog