Merck
  • The comparison of the efficacy and safety of cetirizine, oxatomide, ketotifen, and a placebo for the treatment of childhood perennial allergic rhinitis.
The comparison of the efficacy and safety of cetirizine, oxatomide, ketotifen, and a placebo for the treatment of childhood perennial allergic rhinitis.
Annals of allergy, asthma & immunology : official publication of the American College of Allergy, Asthma, & Immunology (2002-12-19)
Dong-Shang Lai, Ko-Huang Lue, Jie-Cheng Hsieh, Ker-Liang Lin, Hong-Sen Lee
RESUMO
There has been no study comparing the long-term effects of ketotifen, oxatomide, and cetirizine for the treatment of perennial allergic rhinitis among children. We conducted a study to compare the efficacy of the three agents for the treatment of perennial allergic rhinitis among children. The study consisted of a double-blind, placebo-controlled, randomized design, comprising 69 perennial allergic rhinitis patients with mite allergy, aged 6 to 12 years, randomly assigned to 1 of 4 test treatment groups for 3 months: 19 in the cetirizine group (10 mg daily), 18 in the ketotifen group (1 mg, twice daily), 16 in the oxatomide group (1 mg/kg, twice daily), and 16 in the placebo group. We used the nasal symptom score of diary card and the Pediatric Rhinoconjunctivitis Quality of Life Questionnaire and eosinophil cation protein peripheral blood total eosinophil count and immunoglobulin E level, eosinophil proportion from a nasal smear, and nasal peak expiratory flow rate to evaluate the effect of the four agents. Cetirizine was significantly more effective at reducing the mean rhinorrhea score compared with oxatomide for both weeks 8 and 12 (P < 0.01). Before the end of week 12, cetirizine was significantly more effective than ketotifen (P < 0.01). Cetirizine and oxatomide significantly decreased the mean Pediatric Rhinoconjunctivitis Quality of Life Questionnaire score compared with the placebo for week 12 (P < 0.05). Cetirizine was more effective than oxatomide and ketotifen for the relief of nasal congestion and rhinorrhea, and was responsible for significantly decreasing the eosinophil representation of a posttreatment nasal smear.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Oxatomide, ≥99%