Merck
  • Targeting a nuclear anthranilate synthase alpha-subunit gene to the tobacco plastid genome results in enhanced tryptophan biosynthesis. Return of a gene to its pre-endosymbiotic origin.
Targeting a nuclear anthranilate synthase alpha-subunit gene to the tobacco plastid genome results in enhanced tryptophan biosynthesis. Return of a gene to its pre-endosymbiotic origin.
Plant physiology (2001-09-13)
X H Zhang, J E Brotherton, J M Widholm, A R Portis
RESUMO
Anthranilate synthase (AS), the control enzyme of the tryptophan (Trp) biosynthetic pathway, is encoded by nuclear genes, but is transported into the plastids. A tobacco (Nicotiana tabacum) cDNA (ASA2) encoding a feedback-insensitive tobacco AS alpha-subunit was transformed into two different sites of the tobacco plastid genome through site-specific insertion to obtain transplastomic plants with normal phenotype and fertility. A high and uniform level of ASA2 mRNA was observed in the transplastomic plants but not in the wild type. Although the plants with the transgene insertion at ndhF-trnL only expressed one size of the ASA2 mRNA, the plants with the transgene incorporated into the region between accD and open reading frame (ORF) 184 exhibited two species of mRNA, apparently due to readthrough. The transplastomic plants exhibited a higher level of AS alpha-subunit protein and AS enzyme activity that was less sensitive to Trp-feedback inhibition, leading to greatly increased free Trp levels in leaves and total Trp levels in seeds. Resistance to an AS inhibitor, 5-methyl-Trp, was found during seed germination and in suspension cultures of the transplastomic plants. The resistance to the selection agent spectinomycin and to 5-methyl-Trp was transmitted maternally. These results demonstrate the feasibility of modifying the biosynthetic pathways of important metabolites through transformation of the plastid genome by relocating a native gene from the nucleus to the plastid genome. Very high and uniform levels of gene expression can be observed in different lines, probably due to the identical insertion sites, in contrast to nuclear transformation where random insertions occur.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
5-Methyl-DL-tryptophan, tryptophan analog