Merck
Catechol derivatives of aminopyrazine and cell protection against UVB-induced mortality.
Bioorganic & medicinal chemistry (2001-05-17)
J F Cavalier, M Burton, F Dussart, C Marchand, J F Rees, J Marchand-Brynaert
RESUMO
A series of 5-aryl- and 3,5-bis-aryl-2-amino-1,4-pyrazine derivatives 4 and 6, and related imidazolopyrazinones 7, has been synthesized, the aryl groups of which are catechol and/or phenol substituents. These compounds, tested against human keratinocyte cells stressed by UVB irradiation, showed high antioxidative properties. One compound (6f) was more active than EGCG/ECG (green tea extract) in reducing cell mortality.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Aminopyrazine, 98%