Merck
  • [GC-MS analysis of constituents of essential oils from stems of Ephedra sinica Stapf, E. intermedia Schrenk et C.A. Mey. and E. equisetina Bge].
[GC-MS analysis of constituents of essential oils from stems of Ephedra sinica Stapf, E. intermedia Schrenk et C.A. Mey. and E. equisetina Bge].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (1997-08-01)
L Ji, Z Xu, G Pan, G Yang
RESUMO
The essential oils from the dried stems of Ephedra sinica, E. intermedia and E. equisetina were analyzed by GC-MS qualitatively and GC quantitatively. One hundred and twenty-seven constituents were identified, l-alpha-terpineol (31.64%) in E. sinica, 1,4-cineole (12.80%) in E. intermedia and hexadecanoic acid (26.22%) in E. equisetina being the main constituents.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
1,4-Cineole, technical, mixture of isomers, ≥85% (GC)
Supelco
1,4-Cineole, analytical standard