Merck
Synthesis and analgesic activity of N,N'-dicarbonyltryptamines.
Preparative biochemistry & biotechnology (2000-08-05)
H Dongmei, W Chao, Z Ming, P Shiqi
RESUMO
5-Methoxytryptamine and L-tryptophan methyl ester were acylated with malonic acid, dimethyl malonate, or succinic anhydride to produce the corresponding N,N'-dicarbonyltryptamine derivatives. The analgesic activity was evaluated by the tail flick test. All of the compounds exhibited desirable analgesic potency. This result is consistent with that of N-(N-acetyl-L-tryptophanyl)-5-methoxytryptamine and confirmed that introducing substituted tryptamine into the amide chain of melatonin does enhance analgesic potency.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Dimethyl malonate, 98%
Preço e disponibilidade não estão disponíveis no momento.
Sigma-Aldrich
Dimethyl malonate, purum, ≥96.0% (GC)
Preço e disponibilidade não estão disponíveis no momento.