Merck
  • The metal function in the reactions of bovine serum amine oxidase with substrates and hydrazine inhibitors.
The metal function in the reactions of bovine serum amine oxidase with substrates and hydrazine inhibitors.
Journal of biological inorganic chemistry : JBIC : a publication of the Society of Biological Inorganic Chemistry (1999-08-10)
G De Matteis, E Agostinelli, B Mondovì, L Morpurgo
RESUMO
Bovine serum amine oxidase (BSAO) reacts with 2-hydrazinopyridine, which binds the organic co-factor 2,4,5-trihydroxyphenylalanine quinone, forming a band at 435 nm. The band shifts to 526 nm around 60 degrees C, to 415 nm upon denaturation, but only shifts to 429 nm upon Cu2+ depletion. Its wavelength and intensity suggest that the adduct has the azo conformation, whilst the same adduct of crystalline Escherichia coli amine oxidase (ECAO) shows the hydrazone conformation in the X-ray structure. The steady state kinetics of aminomethyl- and aminoethylpyridines confirm that the formation of the product Schiff base, analogous to the azo form of the 2-hydrazinopyridine adduct, is not hindered in solution. The structural stability of the adduct in the absence of Cu2+ is taken to imply hydrogen bonding of the pyridyl nitrogen to a conserved aspartate, as in the ECAO adduct. Thus the ECAO adduct provides a good model for a transient intermediate leading to formation of the BSAO azo adduct. On the basis of this model and of the catalytic competence of Co(2+)-substituted BSAO, confirmed by the present data, a catalytic reaction scheme is proposed.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
2-Hydrazinopyridine, 97%