Merck
The syntaxin family of vesicular transport receptors.
Cell (1993-09-10)
M K Bennett, J E García-Arrarás, L A Elferink, K Peterson, A M Fleming, C D Hazuka, R H Scheller
RESUMO
Syntaxins A and B are nervous system-specific proteins implicated in the docking of synaptic vesicles with the presynaptic plasma membrane. A family of syntaxin-related proteins from rat has been identified that shares 23%-84% amino acid identity. Each of the six syntaxins terminate with a carboxy-terminal hydrophobic domain that anchors the protein on the cytoplasmic surface of cellular membranes. The syntaxins display a broad tissue distribution and, when expressed in COS cells, are targeted to different subcellular compartments. Microinjection studies suggest that the nervous system-specific syntaxin 1A is important for calcium-regulated secretion from neuro-endocrine PC12 cells. These results indicate that the syntaxins are a family of receptors for intracellular transport vesicles and that each target membrane may be identified by a specific member of the syntaxin family.