- Structure of the inhibitor complex of old yellow enzyme from Trypanosoma cruzi.
Structure of the inhibitor complex of old yellow enzyme from Trypanosoma cruzi.
Journal of synchrotron radiation (2010-12-21)
Keishi Yamaguchi, Naoki Okamoto, Keiji Tokuoka, Shigeru Sugiyama, Nahoko Uchiyama, Hiroyoshi Matsumura, Koji Inaka, Yoshihiro Urade, Tsuyoshi Inoue
PMID21169695
RESUMO
Old yellow enzyme (OYE) is an NADPH oxidoreductase which contains flavin mononucleotide as prosthetic group. The X-ray structures of OYE from Trypanosoma cruzi (TcOYE) which produces prostaglandin (PG) F(2α) from PGH(2) have been determined in the presence or absence of menadione. The binding motif of menadione, known as one of the inhibitors for TcOYE, should accelerate the structure-based development of novel anti-chagasic drugs that inhibit PGF(2α) production specifically.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Prostaglandin F Synthase human, recombinant, expressed in E. coli, ≥90% (SDS-PAGE)