Merck
N-linked oligosaccharides predominantly distribute in rat cerebellar cortex.
Okajimas folia anatomica Japonica (2002-01-05)
H Miyata, H Kawakami, T Hori, H Hirano
RESUMO
The distribution or localization of glycoconjugates in rat cerebellar cortex was investigated with 26 different kinds of lectins observed by light and electron microscopy. In paraffin-embedded tissues, PHA-L, PHA-E, DSA, WGA, ConA, LEA, LCA, PSA, and RCA-I, which mainly recognize N-linked oligosaccharide sugar structures, stained the cerebellar cortex, especially the molecular layer. PHA-L staining showed the highest selectivity for the molecular layer among these lectins. Pretreatment with N-glycanase altered the staining intensities of these lectins, whereas pretreatment with O-glycanase did not alter the intensity. In electron microscopy, the cell membrane and Golgi membranes of Purkinje cell, parallel fibers, and synaptic vesicles exhibited a positive reaction with PHA-L. Nuclear pores and synaptic vesicles were positive for WGA binding. These results suggest that there exist N-glycoside binding oligosaccharides predominantly in the cerebellar cortex, especially in the molecular layer, which sugar chains may be relevant to the synaptic transmission in the molecular layer.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
O-Glycosidase from Streptococcus pneumoniae, recombinant, expressed in E. coli, buffered aqueous solution