Merck
Functionalization of reduced graphite oxide sheets with a zwitterionic surfactant.
Chemphyschem : a European journal of chemical physics and physical chemistry (2012-08-15)
Beatriz Martín-García, M Mercedes Velázquez, Francesco Rossella, Vittorio Bellani, Enrique Diez, Jose Luis García Fierro, Jose Antonio Pérez-Hernández, Juan Hernández-Toro, Sergi Claramunt, Albert Cirera
RESUMO
Films of a few layers in thickness of reduced graphite oxide (RGO) sheets functionalized by the zwitterionic surfactant N-dodecyl-N,N-dimethyl-3-ammonio-1-propanesulfonate (DDPS) are obtained by using the Langmuir-Blodgett method. The quality of the RGO sheets is checked by analyzing the degrees of reduction and defect repair by means of X-ray photoelectron spectroscopy, atomic force microscopy (AFM), field-emission scanning electron microscopy (SEM), micro-Raman spectroscopy, and electrical conductivity measurements. A modified Hummers method is used to obtain highly oxidized graphite oxide (GO) together with a centrifugation-based method to improve the quality of GO. The GO samples are reduced by hydrazine or vitamin C. Functionalization of RGO with the zwitterionic surfactant improves the degrees of reduction and defect repair of the two reducing agents and significantly increases the electrical conductivity of paperlike films compared with those prepared from unfunctionalized RGO.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
N-Dodecyl-N,N-dimethyl-3-ammonio-1-propanesulfonate, ≥97.0% (dried material, CHN)
Sigma-Aldrich
N-Dodecyl-N,N-dimethyl-3-ammonio-1-propanesulfonate