Merck
HVEM signalling promotes colitis.
PloS one (2011-05-03)
Corinne Schaer, Stefanie Hiltbrunner, Bettina Ernst, Christoph Mueller, Michael Kurrer, Manfred Kopf, Nicola L Harris
RESUMO
Tumor necrosis factor super family (TNFSF) members regulate important processes involved in cell proliferation, survival and differentiation and are therefore crucial for the balance between homeostasis and inflammatory responses. Several members of the TNFSF are closely associated with inflammatory bowel disease (IBD). Thus, they represent interesting new targets for therapeutic treatment of IBD. We have used mice deficient in TNFSF member HVEM in experimental models of IBD to investigate its role in the disease process. Two models of IBD were employed: i) chemical-induced colitis primarily mediated by innate immune cells; and ii) colitis initiated by CD4(+)CD45RB(high) T cells following their transfer into immuno-deficient RAG1(-/-) hosts. In both models of disease the absence of HVEM resulted in a significant reduction in colitis and inflammatory cytokine production. These data show that HVEM stimulatory signals promote experimental colitis driven by innate or adaptive immune cells.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
CD270 human, recombinant, expressed in E. coli, 0.5 mg protein/mL