Merck
Polyethylene glycol conjugation of recombinant methioninase for cancer therapy.
Protein expression and purification (1998-03-14)
Y Tan, X Sun, M Xu, Z An, X Tan, Q Han, D A Miljkovic, M Yang, R M Hoffman
RESUMO
Recombinant methioninase (rMETase) is a homotetrameric pyridoxal 5'-phosphate enzyme of 172-kda molecular mass derived from Pseudomonas putida and cloned in Escherichia coli. rMETase has been found previously to be an effective, anti-tumor agent in vitro and in vivo. The enzyme targets the elevated minimal methionine requirement seen in all tumor types. In order to prevent immunological reactions which might be produced by multiple dosing of rMETase and to prolong the serum half-life of rMETase, the N-hydroxysuccinimidyl ester of methoxypolyethylene glycol propionic acid (M-SPA-PEG 5000) has been coupled to rMETase. Molar ratios of M-SPA-PEG-5000 (PEG) to rMETase from 10 to 40 were used for PEGylation of rMETase. PEGylation reactions were run at 20 degrees C for 30 to 60 min in reaction buffer (20 mM sodium phosphate buffer, pH 8.3). The PEGylated molecules (PEG-rMETase) were purified from unreacted PEG with Amicon 30 K centriprep concentrators or by Sephacryl S-300 HR gel-filtration chromatography. Unreacted rMETase was removed by DEAE Sepharose FF anion-exchange chromatography. The resulting PEG-rMETase subunit, from a PEG/rMETase ratio of 30/1 in the synthetic reaction, had a molecular mass of approximately 53 kda determined by matrix-assisted laser desorption/ionization mass spectrometry, indicating the conjugation of two PEG molecules per subunit of rMETase and eight per tetramer. PEG-rMETase molecules obtained from reacting ratios of PEG /rMETase of 30/1 had enzyme activities of 70% of unmodified rMETase. PEGylation of rMETase increased the serum half-life of the enzyme in rats to approximately 160 min compared to 80 min for unmodified rMETase. PEG-rMETase could deplete serum methionine levels to less than 0.1 microM for approximately 8 h compared to 2 h for rMETase in rats. Efficacy studies of PEG-rMETase on human lung cancer and kidney cancer cells in vitro demonstrated a 50% inhibitory concentration (IC50) of 0.04 and 0.06 units/ml, respectively. These IC50 values were almost identical to unmodified rMETase, thus indicating maintenance of antitumor efficacy in the PEGylated enzyme. PEG-rMETase had an IC50 for normal lung and kidney cells of 0.8 and 1.5 units/ml, respectively, similar to rMETase. The efficacy data indicated that PEG-rMETase maintained the high level tumor selectivity of rMETase. PEG-rMETase injected intravenously in mice demonstrated a tumor/blood retention ratio of approximately 1/6 compared to 1/10 of unmodified enzyme, indicating that PEG-rMETase distributes to the tumor at least as effectively as rMETase.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Methoxypolyethylene glycol 5,000 propionic acid, ≥80%