- The receptor for the cytotoxic ligand TRAIL.
The receptor for the cytotoxic ligand TRAIL.
Science (New York, N.Y.) (1997-04-04)
G Pan, K O'Rourke, A M Chinnaiyan, R Gentz, R Ebner, J Ni, V M Dixit
PMID9082980
RESUMO
TRAIL (also known as Apo-2L) is a member of the tumor necrosis factor (TNF) ligand family that rapidly induces apoptosis in a variety of transformed cell lines. The human receptor for TRAIL was found to be an undescribed member of the TNF-receptor family (designated death receptor-4, DR4) that contains a cytoplasmic "death domain" capable of engaging the cell suicide apparatus but not the nuclear factor kappa B pathway in the system studied. Unlike Fas, TNFR-1, and DR3, DR4 could not use FADD to transmit the death signal, suggesting the use of distinct proximal signaling machinery. Thus, the DR4-TRAIL axis defines another receptor-ligand pair involved in regulating cell suicide and tissue homeostasis.