Merck
  • Effects of the Deacetylation Degree of Chitosan on 2-Amino-1-methyl-6-phenylimidazo[4,5-b]pyridine (PhIP) Formation in Chemical Models and Beef Patties.
Effects of the Deacetylation Degree of Chitosan on 2-Amino-1-methyl-6-phenylimidazo[4,5-b]pyridine (PhIP) Formation in Chemical Models and Beef Patties.
Journal of agricultural and food chemistry (2021-11-11)
Yueliang Zhao, Hongmei Yang, Nana Zhang, Qian Zhou, Daming Fan, Mingfu Wang
RESUMO
The effects of the deacetylation degree (DD) of chitosan on heterocyclic aromatic amine formation were investigated in chemical models and beef patties. The results in model systems showed that at lower addition levels (10 mg), chitosan with 85% DD showed the strongest inhibitory effect against 2-amino-1-methyl-6-phenylimidazo[4,5-b]pyridine (PhIP) formation, while chitosan with a higher DD (95%) or a lower DD (72 and 50%) did not show any significantly inhibitory effect. Further mechanism study showed that chitosan addition reduced the content of Maillard reaction intermediates including phenylacetaldehyde and the aldol condensation product but increased the PhIP precursor creatinine residue in the chemical model, indicating that chitosan at least partially competed with creatinine to react with phenylacetaldehyde to inhibit PhIP formation. In roast beef patties, 0.15% (w/w) chitosan (85% DD) significantly reduced the formation of PhIP, MeIQx, 4,8-DiMeIQx, Harman, and Norharman by 56.21, 33.32, 31.35, 25.14, and 28.12%, respectively. Moreover, chitosan significantly inhibited the formation of aldehydes in roast beef patties, further confirming the above-mentioned inhibition mechanism. However, the addition of chitosan might promote fatty acid oxidation. In addition, chitosan addition below 0.15% (w/w) had no significant effect on the textural properties of the roast samples.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
PhIP, ≥95% (HPLC)