Merck
Ligand-directed tosyl chemistry for selective native protein labeling in vitro, in cells, and in vivo.
Methods in molecular biology (Clifton, N.J.) (2015-01-07)
Shinya Tsukiji, Itaru Hamachi
RESUMO
Introducing nongenetically encoded, synthetic probes into specific proteins is now recognized as a key component in chemical biology. In particular, the ability to chemically modify specific "native" proteins in various contexts from in vitro to cellular systems is of fundamental importance to study biological systems. We developed a protein-labeling technique termed ligand-directed tosyl (LDT) chemistry for this purpose. This method is capable of labeling specific native proteins with diverse synthetic probes with high site specificity and target selectivity without compromising protein function. Here we describe the principle of the LDT chemistry and the protocol for selective chemical labeling of native carbonic anhydrase in vitro, in blood cells (ex vivo), and in living mice (in vivo).
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Carbonic Anhydrase Isozyme II human, ≥80%, powder, ≥3,000 W-A units/mg protein