Merck
C9orf16 represents the aberrant genetic programs and drives the progression of PDAC.
BMC cancer (2022-10-30)
Xiaojun Chen, Hong Zhang, Bo Xiao
RESUMO
Pancreatic ductal adenocarcinoma (PDAC), constituting 90% of pancreatic cancers, is the fourth leading cause of cancer-related deaths in the world. Lack of early detection of PDAC contributes to its poor prognosis as patients are often diagnosed at an advanced stage of disease. This is mostly due to the lack of promising diagnostic and therapeutic targets and corresponding drugs. Here, by bioinformatic analysis of single cell RNA-sequencing data on normal pancreas tissues, primary and metastatic PDAC tumors, we identified a promising PDAC biomarker, C9orf16. The expression of C9orf16, rarely detectable in normal epithelial cells, was upregulated in primary PDAC cancer cells and was further elevated in metastatic PDAC cancer cells. Gain or loss of function of C9orf16 demonstrated its critical functions in regulating the cell proliferation, invasion and chemotherapy resistance of cancer cells. Pathway analysis and functional studies identified MYC signaling pathways as the most activated pathways in regulating C9orf16 expression and in mediating the development and progression of PDAC. These data suggested a crucial gene regulation system, MYC-C9orf16, which is actively involved in PDAC development and progression, and targeting this system should be a novel diagnostic and therapeutic target for PDAC.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Anti-C9orf16 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution