Merck
Loss of function mutations in ATP2A2 and psychoses: A case report and literature survey.
Psychiatry and clinical neurosciences (2016-04-24)
Takumi Nakamura, An-A Kazuno, Kazuo Nakajima, Ichiro Kusumi, Takashi Tsuboi, Tadafumi Kato
RESUMO
Though genetic factors play a major role in the pathophysiology of psychoses including bipolar disorder (BD) and schizophrenia, lack of well-established causative genetic mutations hampers their neurobiological studies. Darier's disease, an autosomal dominant skin disorder caused by mutations of ATP2A2 on chromosome 12q23-24.1, encoding sarco/endoplasmic reticulum calcium transporting ATPase 2 (SERCA2), reportedly cosegregates with BD. A recent genome-wide association study showed an association of schizophrenia with ATP2A2. We sequenced all coding regions of ATP2A2 in a newly identified patient with Darier's disease and BD. In addition, we performed a literature survey to examine whether likely gene disrupting (LGD) mutations are related to psychoses. We identified a rare heterozygous mutation, c.1288-6A>G, at the 3' end of intron 10 in the patient. A minigene splicing assay showed that this mutation introduces a new splice site causing a frameshift and premature stop codon. A literature survey of case reports of patients with Darier's disease and psychoses revealed that the rate of LGD mutations causing frameshift, altered splicing, gain of stop codon, or loss of start codon was significantly higher among the mutations harbored by these cases (9 of 11) than that of ATP2A2 mutations for which comorbidity of psychosis was not reported (107 of 237, P = 0.026). The only non-LGD mutation (p.C560R) reported in patients with Darier's disease and BD caused decreased ATP2A2 protein expression. These results suggest that psychoses in Darier's disease may be caused by a pleiotropic effect of loss-of-function mutations of ATP2A2.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Antiβ-actina monoclonal, clone AC-15, ascites fluid