Merck
  • Calcium in sympathetic boutons of rat superior cervical ganglion during facilitation, augmentation and potentiation.
Calcium in sympathetic boutons of rat superior cervical ganglion during facilitation, augmentation and potentiation.
Journal of the autonomic nervous system (1998-11-10)
Y Q Lin, K L Brain, M R Bennett
RESUMO
The sympathetic preganglionic nerve terminals of the rat superior cervical ganglion were loaded with the calcium indicator oregon green 488 BAPTA-1 to measure the change in calcium concentration in the terminal boutons, (delta[Ca2+]b) following short (1 or 5 impulses) and long (200 impulses) trains at 30 Hz. The delta[Ca2+]b after a single action potential or a short train declined in two phases: a fast phase with a time constant of 530+/-30 ms and a moderate phase with a time constant of 4.0+/-0.2 s. The delta[Ca2+]b following a long train eventually declined with a time constant of 127+/-34 s (slow phase). The addition of either omega-agatoxin TK (100 nM), omega-conotoxin GVIA (100 nM) or nifedipine (20 microM) to block P-type, N-type or L-type calcium channels respectively showed that the rise in delta[Ca2+ ]b in boutons was predominantly mediated by an influx of calcium through P-type (53+/-7%) and N-type (46+/-4%) calcium channels. Experiments with caffeine, ryanodine and thapsigargin indicate that intracellular caffeine-sensitive calcium stores have a small but statistically significant effect on the fast and moderate phases. The mitochondrial uncoupler carbonyl cyanide m-chlorophenyl hydrazone (CCCP; 2 microM) significantly decreased the amplitude of the slow phase of delta[Ca2+]b relaxation, and sped its time course, suggesting that mitochondria normally dump calcium during this phase. Adenosine reduced the amplitude of delta[Ca2+]b in response to single action potentials by 30+/-6%, suggesting that adenosine-mediated autoinhibition in these boutons reduces Ca2+ influx. Spontaneous increases in delta[Ca2+]b demonstrated Ca2+ coupling between adjacent boutons. The delta[Ca2+]b kinetics are compared with F2 facilitation, augmentation and post-tetanic potentiation.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
ω-Agatoxin TK, ~98% (HPLC), powder