Merck
PAI-1 is a potential transcriptional silencer that supports bladder cancer cell activity.
Scientific reports (2022-07-17)
Hideki Furuya, Yuka Sasaki, Runpu Chen, Rafael Peres, Kanani Hokutan, Kaoru Murakami, Nari Kim, Owen T M Chan, Ian Pagano, Lars Dyrskjøt, Jørgen B Jensen, Per-Uno Malmstrom, Ulrika Segersten, Yijun Sun, Abolfazl Arab, Hani Goodarzi, Steve Goodison, Charles J Rosser
RESUMO
The extracellular activity of Plasminogen activator inhibitor-1 (PAI-1) is well described, acting as an inhibitor of tissue plasminogen activator and urokinase-type plasminogen activator, impacting fibrinolysis. Recent studies have revealed a pro-tumorigenic role of PAI-1 in human cancers, via the regulation of angiogenesis and tumor cell survival. In this study, immunohistochemical staining of 939 human bladder cancer specimens showed that PAI-1 expression levels correlated with tumor grade, tumor stage and overall survival. The typical subcellular localization of PAI-1 is cytoplasmic, but in approximately a quarter of the cases, PAI-1 was observed to be localized to both the tumor cell cytoplasm and the nucleus. To investigate the potential function of nuclear PAI-1 in tumor biology we applied chromatin immunoprecipitation (ChIP)-sequencing, gene expression profiling, and rapid immunoprecipitation mass spectrometry to a pair of bladder cancer cell lines. ChIP-sequencing revealed that PAI-1 can bind DNA at distal intergenic regions, suggesting a role as a transcriptional coregulator. The downregulation of PAI-1 in bladder cancer cell lines caused the upregulation of numerous genes, and the integration of ChIP-sequence and RNA-sequence data identified 57 candidate genes subject to PAI-1 regulation. Taken together, the data suggest that nuclear PAI-1 can influence gene expression programs and support malignancy.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Anti-SERPINE1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution