Merck
  • Genome-wide CRISPR screen for HSV-1 host factors reveals PAPSS1 contributes to heparan sulfate synthesis.
Genome-wide CRISPR screen for HSV-1 host factors reveals PAPSS1 contributes to heparan sulfate synthesis.
Communications biology (2022-07-20)
Takeshi Suzuki, Yoshitaka Sato, Yusuke Okuno, Fumi Goshima, Tadahisa Mikami, Miki Umeda, Takayuki Murata, Takahiro Watanabe, Koichi Watashi, Takaji Wakita, Hiroshi Kitagawa, Hiroshi Kimura
RESUMO
Herpes simplex virus type 1 (HSV-1) is a ubiquitous pathogen that causes various diseases in humans, ranging from common mucocutaneous lesions to severe life-threatening encephalitis. However, our understanding of the interaction between HSV-1 and human host factors remains incomplete. Here, to identify the host factors for HSV-1 infection, we performed a human genome-wide CRISPR screen using near-haploid HAP1 cells, in which gene knockout (KO) could be efficiently achieved. Along with several already known host factors, we identified 3'-phosphoadenosine 5'-phosphosulfate synthase 1 (PAPSS1) as a host factor for HSV-1 infection. The KO of PAPSS1 in HAP1 cells reduced heparan sulfate (HepS) expression, consequently diminishing the binding of HSV-1 and several other HepS-dependent viruses (such as HSV-2, hepatitis B virus, and a human seasonal coronavirus). Hence, our findings provide further insights into the host factor requirements for HSV-1 infection and HepS biosynthesis.