Merck
  • Immunolocalization of glutaryl-CoA dehydrogenase (GCDH) in adult and embryonic rat brain and peripheral tissues.
Immunolocalization of glutaryl-CoA dehydrogenase (GCDH) in adult and embryonic rat brain and peripheral tissues.
Neuroscience (2016-12-17)
Olivier Braissant, Paris Jafari, Noémie Remacle, Hong-Phuc Cudré-Cung, Sonia Do Vale Pereira, Diana Ballhausen
RESUMO
Glutaryl-CoA dehydrogenase (GCDH) is a mitochondrial enzyme that is involved in the degradation of tryptophan, lysine and hydroxylysine. Deficient enzyme activity leads to glutaric aciduria type-I (GA-I). This neurometabolic disease usually manifests with acute encephalopathic crises and striatal neuronal death in early childhood leading to an irreversible dystonic-dyskinetic movement disorder. Fronto-temporal atrophy and white matter changes are already present in the pre-symptomatic period. No detailed information on GCDH expression during embryonic development and in adulthood was available so far. Using immunofluorescence microscopy and cell-type-specific markers to localize GCDH in different tissues, we describe the differential cellular localization of GCDH in adult rat brain and peripheral organs as well as its spatiotemporal expression pattern. During embryonic development GCDH was predominantly expressed in neurons of the central and peripheral nervous system. Significant expression levels were found in epithelial cells (skin, intestinal and nasal mucosa) of rat embryos at different developmental stages. Besides the expected strong expression in liver, GCDH was found to be significantly expressed in neurons of different brain regions, renal proximal tubules, intestinal mucosa and peripheral nerves of adult rats. GCDH was found widely expressed in embryonic and adult rat tissues. In rat embryos GCDH is predominantly expressed in brain implying an important role for brain development. Interestingly, GCDH was found to be significantly expressed in different other organs (e.g. kidney, gut) in adult rats probably explaining the evolving phenotype in GA-I patients.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Anti-GCDH antibody produced in rabbit, IgG fraction of antiserum